2,4,6-Tribromophenol 98%
:118-79-6
:
:C₆H₃Br₃O
:MFCD00002150
:290
:330.8 g/mol
:89…93 °C
:12,09744
:204-278-6
:3077
:GHS07
:GHS09
:Ambient
- Environmentally Preferable:
- Appearance:White to pink to light brown Powder or flakes and/or chunks
- Infrared spectrum:Conforms
- Melting point:89 to 94 °C
- Titration with NaOH:≥97.5 %
Frequently Bought Together