4-Bromoanisole 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:104-92-7
:
Formula:C₇H₇BrO
MDL Number:MFCD00000097
:187.04 g/mol
Merck Index:13,01412
Boiling Pt:223 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1.49 g/cm³
Flash Pt:94 °C
Storage Temperature:Ambient
EINECS:203-252-1
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5630 to 1.5650 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together