1-Bromo-2-nitrobenzene ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Bromonitrobenzene
CAS Number:577-19-5
:
Formula:C₆H₄BrNO₂
MDL Number:MFCD00007045
Boiling Pt:261 °C
Melting Pt:40…44 °C
Density:1.62 g/cm³ (20 °C)
Molecular Weight:202.01 g/mol
Flash Pt:112 °C (233 °F)
Storage Temperature:Ambient
EINECS:209-409-0
UN:3459
ADR:6.1 III
:GHS06
:GHS08
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together