

4-Bromoaniline 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-40-1
:
Formula:C₆H₆BrN
MDL Number:MFCD00007822
Merck Index:13,01388
Boiling Pt:230…250 °C
Melting Pt:60…64 °C
Density:1.497 g/cm³
Flash Pt:>110 °C
UN:2811
ADR:6.1,III
:172.02 g/mol
EINECS:203-393-9
:GHS06
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to grey-brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:60 to 64 °C
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together