Dithiooxamide, Supelco®
Catalog # 1.00629.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Solubility:0.37 g/l (20 °C)
- Melting Point:200 °C (decomposition)
- Molar Mass:120.20 g/mol
- Bulk Density:650 kg/m3
Specification Test Results
About this item
:
:120.2 g/mol
:M-Clarity™ quality level = MQ100
EINECS:201-203-9
:GHS07
CAS Number:79-40-3
Formula:C₂H₄N₂S₂
MDL Number:MFCD00004941