Pesticide reference standard, Hexachlorobenzene 100 µg/ml in methanol, Dr Ehrenstorfer
EHERXA14160000ME
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:118-74-1
- Description:Hexachlorobenzene 100 µg/ml in methanol
- Formula:C₆Cl₆
- Description:Hexachlorobenzene