Order Entry
United Kingdom
ContactUsLinkComponent
Organic reference standard, 2-Phenylnaphthalene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Organic reference standard, 2-Phenylnaphthalene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20923100CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2-Phenylnaphthalene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20923100CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    612-94-2
  • Description:
    2-Phenylnaphthalene 10 µg/ml in cyclohexane
  • Formula:
    C₁₆H₁₂
  • Description:
    2-Phenylnaphthalene

Specifications

About this item