Order Entry
United Kingdom
ContactUsLinkComponent
Organic reference standard, Aniline 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Organic reference standard, Aniline 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL10262500CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Aniline 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL10262500CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    62-53-3
  • Description:
    Aniline 10 µg/ml in cyclohexane
  • Formula:
    C₆H₇N
  • Description:
    Aniline

Specifications

About this item