Order Entry
United Kingdom
ContactUsLinkComponent
2,4,6-Triallyloxy-1,3,5-triazine ≥98% stabilised
Toxic
Irritant
2,4,6-Triallyloxy-1,3,5-triazine ≥98% stabilised
Catalog # L16274.18
CAS Number:  
Toxic
Irritant
2,4,6-Triallyloxy-1,3,5-triazine ≥98% stabilised
Catalog # L16274.18
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

CAS Number:101-37-1
 :
Formula:C₁₂H₁₅N₃O₃
 :249.27 g/mol
Boiling Pt:149…150 °C (4 mmHg)
Melting Pt:25…28 °C
Density:1.11
Flash Pt:166 °C (331 °F)
Storage Temperature:Ambient
EINECS:202-936-7
ADR:9,III
 :Stabilised with hydroquinone (HQ)
UN:3077
 :GHS06
 :GHS07
MDL Number:MFCD00006049