Sodium borohydride, powder
Catalog # ABCRAB101969.0500
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:37.83 g/mol
Merck Index:13,08664
Melting Pt:400 °C
Density:1.07 g/cm³ (20 °C)
Flash Pt:69 °C
Storage Temperature:Ambient
EINECS:241-004-4
UN:1426
ADR:4.3,I
:GHS02
:GHS06
:GHS05
CAS Number:16940-66-2
Formula:NaBH₄
MDL Number:MFCD00003518