4-Bromo-2-chlorophenol ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 3964-56-5 Molecular Formula: C6H4BrClO Formula Weight: 207.46 Flash Point: 110°(230°F) Odour: Phenol-like Boiling Point: 232-235° Beilstein Reference: 2042867 MDL No.: MFCD00002166
- CAS Number:3964-56-5
- Formula:C₆H₄BrClO
- MDL Number:MFCD00002166
- Molecular Weight:207.45 g/mol
- Boiling Pt:232…235 °C
- Melting Pt:47…50 °C
- Density:1.617 g/cm³ (20 °C)
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:223-572-5
- UN:3082
- ADR:9,III
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 3964-56-5 Molecular Formula: C6H4BrClO Formula Weight: 207.46 Flash Point: 110°(230°F) Odour: Phenol-like Boiling Point: 232-235° Beilstein Reference: 2042867 MDL No.: MFCD00002166
- CAS Number:3964-56-5
- Formula:C₆H₄BrClO
- MDL Number:MFCD00002166
- Molecular Weight:207.45 g/mol
- Boiling Pt:232…235 °C
- Melting Pt:47…50 °C
- Density:1.617 g/cm³ (20 °C)
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:223-572-5
- UN:3082
- ADR:9,III



