Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2,4-Tribromobenzene
1,2,4-Tribromobenzene
1,2,4-Tribromobenzene
1,2,4-Tribromobenzene
EHERC17664900
:
EHRENSTORFER
:
1,2,4-Tribromobenzene
EHERC17664900
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,1 g
Seq:
3020
:
:
314.8 g/mol
:
85…90 °C (0.04 mmHg)
:
85…90 °C (0,04 mmHg)
:
44 °C
:
210-433-9
:
GHS07
:
615-54-3
:
C₆H₃Br₃
:
MFCD00000062