Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Methylanthracene
1-Methylanthracene
1-Methylanthracene
EHERC20834900
:
EHRENSTORFER
:
1-Methylanthracene
EHERC20834900
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
10 mg
Seq:
2275
:
:
192.26 g/mol
:
363 °C (1013 hPa)
:
86…88 °C
:
1.11 g/cm³ (20 °C)
:
210-224-2
:
610-48-0
:
C₁₅H₁₂