


2-Methyl-2-butene ≥95% (by GC), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:513-35-9
:26760-64-5
Formula:C₅H₁₀
MDL Number:MFCD00009276
:70.13 g/mol
Merck Index:12,00649
Boiling Pt:35…38 °C (lit.)
Melting Pt:–124 °C
Density:0.6623 g/cm³ (20 °C)
Flash Pt:–45 °C
Storage Temperature:Refrigerator
EINECS:208-156-3
UN:2460
ADR:3,II
:GHS02
:GHS08
:GHS07
Frequently Bought Together