Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
1-Docosene
1-Docosene
1-Docosene
Print
1-Docosene
Catalog #
1-Docosene
Catalog #
Supplier:
CAS Number:
1599-67-3
Expand image
Return to Top
1-Docosene
Add to cart
CAS Number:
1599-67-3
Return Policy
Product Details & Documents
CAS Number:
1599-67-3
Formula:
C₂₂H₄₄
MDL Number:
MFCD00027085
Molecular Weight:
308.59 g/mol
Boiling Pt:
162 °C (0,1 mmHg)
Melting Pt:
58…60 °C
Density:
0,8002 g/cm³ (20 °C)
EINECS:
216-490-6