1H,1H,2H-Perfluoro-1-decene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:21652-58-4
:
Formula:C₁₀H₃F₁₇
MDL Number:MFCD00039246
:446.11 g/mol
Boiling Pt:145…150 °C (1013 hPa)
Density:1.65 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:244-503-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together