0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,3,3-Trifluoropropene
3,3,3-Trifluoropropene
3,3,3-Trifluoropropene
ABCRAB101410.0100
:
abcr
:
3,3,3-Trifluoropropene
ABCRAB101410.0100
:
abcr
:
Pack type:
Glass bottle for solids
Pk:
100 g
:
:
96.05 g/mol
:
–18 °C (1013 hPa)
:
0.94 g/cm³ (20 °C)
:
211-637-0
:
1954
:
2,
:
677-21-4
:
C₃H₃F₃
:
MFCD00039265