Order Entry
United Kingdom
ContactUsLinkComponent
4-Bromo-1-butene 98+%
4-Bromo-1-butene 98+%
Certificates
undefined
4-Bromo-1-butene 98+%

Specification Test Results

  • Appearance:
    Clear colorless to light brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4595 to 1.4635 (20°C, 589 nm)
  • GC:
    ≥98.0 %

Specification Test Results

Related Information

Frequently Bought Together