

2-Methyl-3-heptanone
Catalog # EHERC15087900
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:128.21 g/mol
Boiling Pt:158 °C (1013 hPa)
Melting Pt:–59 °C
Density:0.817 g/cm³ (20 °C)
EINECS:235-877-0
UN:1224
ADR:3,III
:GHS02
:GHS07
CAS Number:13019-20-0
Formula:C₈H₁₆O
MDL Number:MFCD00009481