

Sodium dodecyl sulphate (SDS), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:151-21-3
:
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
:288.38 g/mol
Merck Index:14,08636
Melting Pt:204…207 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:>100 °C
UN:1325
ADR:4.1,II
:M-Clarity™ quality level = MQ200
EINECS:205-788-1
:GHS02
:GHS06
Storage Temperature:Ambient
Frequently Bought Together