Order Entry
United Kingdom
ContactUsLinkComponent
Ethylene glycol
Irritant
Ethylene glycol
Catalog # SERA11285.02
Supplier:  Serva
CAS Number:  
undefined
Irritant
Ethylene glycol
Catalog # SERA11285.02
Supplier:  Serva
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :62.07 g/mol
Merck Index:13,03832
Boiling Pt:196…198 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.115 g/cm³ (20 °C)
Flash Pt:116 °C
Storage Temperature:Refrigerator
EINECS:203-473-3
REACH:01-2119456816-28
 :GHS07
CAS Number:107-21-1
Formula:HOCH₂CH₂OH
MDL Number:MFCD00002885