

2-Propanol ≥99.7% for HPLC
22906.K7
:
- Pack type:Glass bottle
- Pk:4 L
- Seq:5162
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
:67-63-0
:
:(CH₃)₂CHOH
:MFCD00011674
:60.1 g/mol
:14,5208
:82…83 °C
:–90 °C
:0.785
:12 °C (53 °F)
:Ambient
:200-661-7
:1219
:3,II
:01-2119457558-25
:GHS02
:GHS07