Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Heptanol
1-Heptanol
1-Heptanol
1-Heptanol
PROOCHE137
:
LGC Standards PROMOCHEM
:
1-Heptanol
PROOCHE137
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
2 mL
Seq:
2145
:
:
116.2 g/mol
:
13,04681
:
176 °C (1013 hPa)
:
–35 °C
:
0.82 g/cm³ (20 °C)
:
70 °C
:
Ambient
:
203-897-9
:
GHS07
:
111-70-6
:
H₃C(CH₂)₅CH₂OH
:
MFCD00002986