Arachidyl alcohol
Catalog # EHERC13113000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
- Environmentally Preferable:
Specifications
About this item
Synonyms: 1-Eicosanol
:
:298.55 g/mol
Boiling Pt:309
Melting Pt:71.5
Density:0.88 g/cm³ (21 °C)
Storage Temperature:Ambient
EINECS:211-119-4
CAS Number:629-96-9
Formula:H₃C(CH₂)₁₈CH₂OH
MDL Number:MFCD00002938