Order Entry
United Kingdom
ContactUsLinkComponent
1-Butanol
  ICNA19400191
 :  
undefined
1-Butanol
  ICNA19400191
 :  
  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
 :
 :74.12 g/mol
 :13,01539
 :117.6 °C (1013 hPa)
 :–89 °C
 :0.812 g/cm³ (20 °C)
 :30 °C
 :Ambient
 :200-751-6
 :1120
 :3,III
 :01-2119484630-38
 :GHS02
 :GHS05
 :GHS07
 :71-36-3
 :H₃C(CH₂)₂CH₂OH
 :MFCD00002964