2,2,3,3-Tetrafluoro-1-propanol
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:76-37-9
:
Formula:C₃H₄F₄O
MDL Number:MFCD00004676
:132.06 g/mol
Merck Index:12,09346
Boiling Pt:109 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.6647 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-955-5
UN:1987
ADR:3,III
Frequently Bought Together