Order Entry
United Kingdom
ContactUsLinkComponent
1-Iodoperfluorbutane 99%
1-Iodoperfluorbutane 99%
Certificates
undefined
1-Iodoperfluorbutane 99%

Specification Test Results

  • Appearance:
    Clear purple Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3275 to 1.3295 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together