

Dichloromethane stabilised, Uvasol® for spectroscopy, Supelco®
Catalog # 1.06048.2500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Solubility:20 g/L (20 °C)
- Melting Point:-95 °C
- Molar Mass:84.93 g/mol
- Boiling Point:40 °C (1013 hPa)
- Vapor Pressure:475 hPa (20 °C)
- Refractive Index:1.42
- Explosion Limit:13 - 22%(V)
- Density:1.33 g/cm³ (20 °C)
- pH:(H₂O, 20 °C) neutral
- Ignition Point:605 °C DIN 51794
- Evaporation Number:1.9
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
About this item
:
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:Stabilised with 2-methyl-2-butene (iso-amylene) 50 ppm
EINECS:200-838-9
UN:1593
:GHS08
:GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881