Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Valeric acid ≥99%

Valeric acid ≥99%

Corrosive
Irritant
Valeric acid ≥99%
Certificates
Valeric acid ≥99%
Valeric acid ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 109-52-4 Molecular Formula: C5H10O2 Formula Weight: 102.13 Flash Point: 88°(190°F) Refractive Index: 1.4080 Odour: Unpleasant Melting Point: -34° Boiling Point: 184-186° Beilstein Reference: 969454 Merck Reference: 14,9904 UN No.: 3265 MDL No.: MFCD00004413

Synonyms:  Pentanoic acid
  • CAS Number:109-52-4
  • Formula:C₅H₁₀O₂
  • MDL Number:MFCD00004413
  • Molecular Weight:102.13 g/mol
  • Merck Index:14,9904
  • Boiling Pt:184…186 °C
  • Melting Pt:–34 °C
  • Density:0.938
  • Flash Pt:88 °C (190 °F)
  • Storage Temperature:Ambient
  • EINECS:203-677-2
  • UN:3265
  • ADR:8,III
Product Family Options

Product Information