

Butyric acid 99+%
:107-92-6
:
:C₄H₈O₂
:MFCD00002814
:13,01593
:162…165 °C (760 mmHg)
:7…5 °C
:0.96 g/cm³
:69 °C
:2820
:8,III
:88.11 g/mol
:203-532-3
:GHS05
:GHS07
:Ambient
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3965 to 1.3995 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥99.0 %
- Color scale:≤15 APHA
Frequently Bought Together