3-Pentanone ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 96-22-0 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 7°(45°F) Refractive Index: 1.3920 Odour: Acetone-like Melting Point: -39° Boiling Point: 102-103° Beilstein Reference: 635749 Merck Reference: 14,3121 UN No.: 1156 MDL No.: MFCD00009320
Synonyms: Diethyl ketone
- CAS Number:96-22-0
- Formula:C₅H₁₀O
- MDL Number:MFCD00009320
- Molecular Weight:86.13 g/mol
- Merck Index:14,3121
- Boiling Pt:102…103 °C
- Melting Pt:–39 °C
- Density:0.812
- Flash Pt:7 °C (45 °F)
- Storage Temperature:Ambient
- EINECS:202-490-3
- UN:1156
- ADR:3,II
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 96-22-0 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 7°(45°F) Refractive Index: 1.3920 Odour: Acetone-like Melting Point: -39° Boiling Point: 102-103° Beilstein Reference: 635749 Merck Reference: 14,3121 UN No.: 1156 MDL No.: MFCD00009320
Synonyms: Diethyl ketone
- CAS Number:96-22-0
- Formula:C₅H₁₀O
- MDL Number:MFCD00009320
- Molecular Weight:86.13 g/mol
- Merck Index:14,3121
- Boiling Pt:102…103 °C
- Melting Pt:–39 °C
- Density:0.812
- Flash Pt:7 °C (45 °F)
- Storage Temperature:Ambient
- EINECS:202-490-3
- UN:1156
- ADR:3,II



