Copper(II) phthalocyanine
Catalog # EHERC16206500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
- Environmentally Preferable:
Specifications
About this item
Synonyms: 29H,31H-Phthalocyaninato-N29,N30,N31,N32 copper, Pigment Blue 15
:
:576.08 g/mol
EINECS:205-685-1
CAS Number:147-14-8
Formula:C₃₂H₁₆CuN₈
MDL Number:MFCD00010719
Melting Pt:480
Merck Index:12,02584