Order Entry
United Kingdom
ContactUsLinkComponent
Water PCR tested for molecular biology DNA-free , suited for qPCR
Water PCR tested for molecular biology DNA-free , suited for qPCR
Catalog # A8510.1017
Supplier:  PanReac AppliChem
CAS Number:  
Water PCR tested for molecular biology DNA-free , suited for qPCR
Catalog # A8510.1017
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075
  • Pk Info:
    10× 1,7 ml

Specifications

Specification Test Results

  • Bacterial DNA:
    not detectable (min. 40 PCR cycles)
  • DNases/RNases:
    not detectable

Specification Test Results

About this item

 :
 :18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332