Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Acetic acid 0,1%

Acetic acid 0,1%

Glass bottle, 1 L
Catalog # ROTHCP04.1
Marketsource
Supplier:   Roth Carl
Acetic acid 0,1%
Catalog # ROTHCP04.1
Supplier:  Roth Carl
Marketsource
CAS Number:  64-19-7
Acetic acid 0,1%
Acetic acid 0,1%

Glass bottle, 1 L

CAS Number:  64-19-7
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Vinegar acid, Methanecarboxylic acid, Ethanoic acid
  • CAS Number:64-19-7
  • Formula:H₃CCOOH
  • MDL Number:MFCD00036152
  • Molecular Weight:60.05 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-580-7
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.