Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Digitoxin

Digitoxin

1 g
Catalog # MOLEM70068568
Marketsource
Supplier:   Molekula
Digitoxin
Catalog # MOLEM70068568
Supplier:  Molekula
Marketsource
CAS Number:  71-63-6
Digitoxin

1 g

CAS Number:  71-63-6
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:71-63-6
  • Formula:C₄₁H₆₄O₁₃
  • MDL Number:MFCD00003686
  • Molecular Weight:764.95 g/mol
  • Melting Pt:276 °C
  • EINECS:200-760-5
  • UN:2811
  • ADR:6.1,II
Specifications

Specifications

  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.