Dexamethasone, Sigma-Aldrich®
Catalog # SIALD8893-1MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 mg
- Seq:2070
- Environmentally Preferable:
Specifications
About this item
Synonyms: Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
:
:392.47 g/mol
EINECS:200-003-9
CAS Number:50-02-2
Formula:C₂₂H₂₉FO₅
MDL Number:MFCD00064136
Melting Pt:268…271 °C
:M-Clarity™ quality level = MQ200