Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
6-Nitroindole

6-Nitroindole

Marketsource
Supplier:   COMBI-BLOCKS
6-Nitroindole
Supplier:  COMBI-BLOCKS
Marketsource
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  6-Nitro-1H-indole
  • CAS Number:4769-96-4
  • Formula:C₈H₆N₂O₂
  • MDL Number:MFCD00051497
  • Molecular Weight:162.15 g/mol
  • EINECS:225-311-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.