Order Entry
United Kingdom
ContactUsLinkComponent
4-Bromo-2,5-difluorobenzenesulphonyl chloride, Sigma-Aldrich®
  SIAL632856-1G
 :  Merck
 :  
4-Bromo-2,5-difluorobenzenesulphonyl chloride, Sigma-Aldrich®
  SIAL632856-1G
 :  Merck
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047
 :
 :291.5 g/mol
 :76 °C (0.01 mmHg)
 :34…36 °C
 :8,II
 :M-Clarity™ quality level = MQ200
 :3261
 :207974-14-9
 :C₆H₂BrClF₂O₂S
 :MFCD00042181