Order Entry
United Kingdom
ContactUsLinkComponent
4-Bromo-2,5-difluorobenzenesulphonyl chloride ≥97%
Corrosive
4-Bromo-2,5-difluorobenzenesulphonyl chloride ≥97%
Catalog # A19634.03
CAS Number:  
Corrosive
4-Bromo-2,5-difluorobenzenesulphonyl chloride ≥97%
Catalog # A19634.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

CAS Number:207974-14-9
 :
Formula:C₆H₂BrClF₂O₂S
MDL Number:MFCD00042181
 :291.5 g/mol
Boiling Pt:76 °C (0.01 mmHg)
Melting Pt:34…36 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05