Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
6,7-Dihydroxyflavone

6,7-Dihydroxyflavone

Glass bottle for solids, 10 mg
Catalog # ROTH8066.1
Marketsource
Supplier:   Roth Carl
6,7-Dihydroxyflavone
Catalog # ROTH8066.1
Supplier:  Roth Carl
Marketsource
CAS Number:  38183-04-9
6,7-Dihydroxyflavone

Glass bottle for solids, 10 mg

CAS Number:  38183-04-9
Price
Inquire For Price
Log in for availability
1 * 10 mg
Product Details & Documents
Synonyms:  6,7-Dihydroxy-2-phenylchromen-4-one, 6,7-Dihydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 6,7-Dihydroxy-2-phenyl-4H-1-benzopyran-4-one, 6,7-Dihydroxy-2-phenyl-4H-chromen-4-one, 6,7-Dihydroxy-2-phenyl-4-benzopyrone
  • CAS Number:38183-04-9
  • Formula:C₁₅H₁₀O₄
  • Molecular Weight:254.24 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.