4-(Aminomethyl)benzoic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-91-7
:
Formula:C₈H₉NO₂
MDL Number:MFCD00010203
Molecular Weight:151.16 g/mol
Melting Pt:>300 °C
EINECS:200-297-9
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Water:≤6 % (K.F.)
- HPLC:≥98.5 %
- Titration with HClO4:98.5 to 101.5 % (On dry substance)
Specification Test Results
Frequently Bought Together