
Naphthol AS 99%
Catalog # ACRO212181000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance:Beige Powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Melting point:≥243°C
- Water:≤0.3 %
- Titration excess NaOH:≥98.5 %
Specification Test Results
About this item
:
Boiling Pt:494 °C (1013 hPa)
Melting Pt:>243 °C
:263.3 g/mol
EINECS:202-188-1
:GHS07
CAS Number:92-77-3
Formula:C₁₇H₁₃NO₂
MDL Number:MFCD00004096