Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Amino-5-bromobenzoic acid
3-Amino-5-bromobenzoic acid, Sigma-Aldrich®
3-Amino-5-bromobenzoic acid, Sigma-Aldrich®
SIAL686425-5G
:
Merck
:
3-Amino-5-bromobenzoic acid, Sigma-Aldrich®
SIAL686425-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
216.03 g/mol
:
6.1,III
:
M-Clarity™ quality level = MQ100
:
2811
:
42237-85-4
:
C₇H₆BrNO₂
:
MFCD00227745