Order Entry
United Kingdom
ContactUsLinkComponent
 
3-(Trifluoromethyl)-1H-pyrazole, Sigma-Aldrich®
Catalog # SIAL406228-5G
Supplier:  Merck
CAS Number:  
3-(Trifluoromethyl)-1H-pyrazole, Sigma-Aldrich®
Catalog # SIAL406228-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3-(Trifluoromethyl)-1H-pyrazole, 3-(Trifluoromethyl)pyrazole
 :
 :136.08 g/mol
 :M-Clarity™ quality level = MQ200
CAS Number:20154-03-4
Formula:C₄H₃F₃N₂
MDL Number:MFCD00115018