2-Bromo-4,6-difluorobenzenesulphonyl chloride
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:351003-42-4
:
Formula:C₆H₂BrClF₂O₂S
MDL Number:MFCD03094561
:291.5 g/mol
Boiling Pt:265…267 °C (1013 hPa)
Density:1.93 g/cm³ (20 °C)
UN:3265
ADR:8,II
Frequently Bought Together