2-Bromo-3-methoxypyridine
Marketsource
Supplier: COMBI-BLOCKS
Jump to:
About this item
- CAS Number:24100-18-3
- Formula:C₆H₆BrNO
- MDL Number:MFCD01570896
- Molecular Weight:188.02 g/mol
- EINECS:246-017-9
2 Options Available Below
Product Details & Documents
- CAS Number:24100-18-3
- Formula:C₆H₆BrNO
- MDL Number:MFCD01570896
- Molecular Weight:188.02 g/mol
- EINECS:246-017-9
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



