Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

4'-Propylacetophenone

1 g
Catalog # FLUO022390-1G
Marketsource
Supplier:   FLUOROCHEM
4'-Propylacetophenone
Catalog # FLUO022390-1G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  2932-65-2
4'-Propylacetophenone

1 g

CAS Number:  2932-65-2
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  1-(4-Propylphenyl)ethanone, p-Propylacetophenone
  • CAS Number:2932-65-2
  • Formula:C₁₁H₁₄O
  • MDL Number:MFCD00041359
  • Molecular Weight:162.23
  • EINECS:220-903-5
Specifications

Specifications

  • Pk:
    1 g

Specifications