2-Bromo-5-chlorophenol
Catalog # COBBOT-0707-1G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:207.45 g/mol
CAS Number:13659-23-9
Formula:C₆H₄BrClO
MDL Number:MFCD00142870