Pesticide reference standard, 3,5-Dichloroaniline 100 µg/ml in methanol, Dr Ehrenstorfer
EHERXA12323500ME
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:626-43-7
- Description:3,5-Dichloroaniline 100 µg/ml in methanol
- Formula:C₆H₅Cl₂N
- Description:3,5-Dichloroaniline