Organic reference standard, 2,4-Xylidine 100 µg/ml in acetonitrile, Dr Ehrenstorfer
EHERXA12724500AL
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:95-68-1
- Description:2,4-Xylidine 100 µg/ml in acetonitrile
- Formula:C₈H₁₁N
- Description:2,4-Xylidine